2-Chlor-3,6-difluorphenol
Lieferant: FLUOROCHEM
Synonyme:
2-Choro-3,6-difluorophenol
005604-5G
005604-1G
FLUO005604-5GEA
321
EUR
FLUO005604-5G
FLUO005604-1G
2-Chlor-3,6-difluorphenol
2-Chlor-3,6-difluorphenol
Formel:
C₆H₃ClF₂O Molecular Weight: 164.54 g/mol |
MDL:
MFCD01631450 CAS-Nummer: 261762-50-9 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...