CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure) ≥98%
Lieferant: Thermo Scientific
Synonyme:
3-(Cyclohexylamino)-2-hydroxy-1-propanesulphonic acid
B21305.14
B21305.22
B21305.36
B21305.14EA
34.8
EUR
B21305.14
B21305.22
B21305.36
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure) ≥98%
CAPSO (3-(Cyclohexylamino)-2-hydroxypropansulfonsäure)
Purity: 98%
CAS No.: 73463-39-5
Molecular Formula: C9H19NO4S
Formula Weight: 237.32
Melting Point: 270-274°
MDL No.: MFCD00041778
Formel:
C₉H₁₉NO₄S Molecular Weight: 237,32 g/mol Schmelzpunkt: 270…274 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041778 CAS-Nummer: 73463-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...