CAPS (3-(Cyclohexylamino)propansulfonsäure), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Cyclohexylaminopropanesulphonic acid

M-Clarity™ Qualitätsstufe = MQ400

C2632-100G C2632-1KG C2632-250G C2632-25G C2632-6X100G
SIALC2632-100GEA 153 EUR
SIALC2632-100G SIALC2632-1KG SIALC2632-250G SIALC2632-25G SIALC2632-6X100G
CAPS (3-(Cyclohexylamino)propansulfonsäure), Sigma-Aldrich®
CAPS (3-(Cyclohexylamino)propansulfonsäure)
Formel: C₆H₁₁NH(CH₂)₃SO₃H
Molecular Weight: 221.32 g/mol
Schmelzpunkt: 324
Lagertemperatur: Raumtemperatur
MDL: MFCD00003837
CAS-Nummer: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR