3,4,5-Trichlor-6-phenylpyridazin

Lieferant: CHEMPUR

BTB12379.250 BTB12379.1
CHMPBTB12379.250EA 0 EUR
CHMPBTB12379.250 MAYBBTB12379.1
3,4,5-Trichlor-6-phenylpyridazin
3,4,5-Trichlor-6-phenylpyridazin
Formel: C₁₀H₅Cl₃N₂
Molecular Weight: 259,52 g/mol
CAS-Nummer: 51448-00-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR