Dihydrocapsaisin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 8-Methyl-N-vanillylnonansäureamid, N-(4-Hydroxy- 3-methoxybenzyl)-8-methylnonansäureamid

M-Clarity™ Qualitätsstufe = MQ200

M1022-1G M1022-250MG M1022-50MG
SIALM1022-1GEA 1140 EUR
SIALM1022-1G SIALM1022-250MG SIALM1022-50MG
Dihydrocapsaisin, Sigma-Aldrich®
Dihydrocapsaisin
Formel: C₁₈H₂₉NO₃
Molecular Weight: 307.43 g/mol
MDL: MFCD00057794
CAS-Nummer: 19408-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR