Dihydrocapsaisin

Lieferant: Cayman Chemical

Synonyme: 8-Methyl-N-vanillylnonansäureamid, N-(4-Hydroxy- 3-methoxybenzyl)-8-methylnonansäureamid

92355-10 92355-100 92355-5 92355-50
CAYM92355-10EA 81 EUR
CAYM92355-10 CAYM92355-100 CAYM92355-5 CAYM92355-50
Dihydrocapsaisin
Dihydrocapsaisin
Formel: C₁₈H₂₉NO₃
Molecular Weight: 307.43 g/mol
MDL: MFCD00057794
CAS-Nummer: 19408-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR