Trityliumhexafluorphosphat ≥98%
Lieferant: Thermo Scientific
Gefahr
Synonyme: Triphenylcarboniumhexafluorophosphat
L03140.09 L03140.18
L03140.09EA 101 EUR
L03140.09 L03140.18
Trityliumhexafluorphosphat ≥98%
Trityliumhexafluorphosphat
Purity: 98% CAS No.: 437-17-2 Molecular Formula: C19H15F6P Formula Weight: 388.29 Storage Temperature: -30°C to -10°C Melting Point: ca 160° dec. Beilstein Reference: 4344297 UN No.: 3261 MDL No.: MFCD00013121
Formel: C₁₉H₁₅F₆P
Molecular Weight: 388,29 g/mol
Schmelzpunkt: ∼ 160 °C (Zers.)
MDL: MFCD00013121
CAS-Nummer: 437-17-2
EINECS: 207-112-0
UN: 3261
ADR: 8,II