Trityliumtetrafluoroborat 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Triphenylcarboniumtetrafluoroborat
172950250
172950050
172951000
ACRO172950250EA
183
EUR
ACRO172950250
ACRO172950050
ACRO172951000
Trityliumtetrafluoroborat 97%
Trityliumtetrafluoroborat
Formel:
C₁₉H₁₅BF₄ Molecular Weight: 330.13 g/mol Schmelzpunkt: 205…215 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00013120 CAS-Nummer: 341-02-6 EINECS: 206-433-3 UN: 3261 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | Yellow to green to brown Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 203 to 213 °C (Dec) |
NMR | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...