Rubidium ≥99,75% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr


Bruchsiegelampulle

10315.03 10315.06 10315.14 10315.22
10315.03EA 165 EUR
10315.03 10315.06 10315.14 10315.22
Rubidium ≥99,75% (Metall-Basis)
Rubidium

Purity: 99.75% CAS No.: 7440-17-7 Physical Form: Vacuum sealed in breakseal ampoules Appearance: Silver-white UN No.: 1423 MDL No.: MFCD00134055

Formel: Rb
Molecular Weight: 85,47 g/mol
Siedepunkt: 700 °C (1013 hPa)
Schmelzpunkt: 39 °C
Dichte: 0,83 g/cm³ (1399.85 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134055
CAS-Nummer: 7440-17-7
EINECS: 231-126-6
UN: 1423
ADR: 4.3,I
Merck Index: 12,08439

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR