Iod ≥99%
Lieferant: Thermo Fisher Scientific
Gefahr
A12278.0B A12278.18 A12278.30
A12278.0BEA 273 EUR
A12278.0B A12278.18 A12278.30
Iod ≥99%
Iod
Purity: 99+% CAS No.: 7553-56-2 Molecular Formula: I2 Formula Weight: 253.81 Flash Point: None Physical Form: Cryst solid Density: 4.930 Appearance: Grey to purple Odour: Irritating Melting Point: 183-186° subl. Beilstein Reference: 969212 Merck Reference: 14,5014 UN No.: 3495 MDL No.: MFCD00011355
Formel: I₂
Molecular Weight: 253.81 g/mol
Siedepunkt: 184.3
Schmelzpunkt: 114
Dichte: 4.9 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011355
CAS-Nummer: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036