Praseodym 99,9 REO basis, foil, Dicke 0.1 mm (0.004 in)

Lieferant: Thermo Fisher Scientific
Gefahr

13962.FF 13962.FI
13962.FFEA 215 EUR
13962.FF 13962.FI
Praseodym 99,9 REO basis, foil, Dicke 0.1 mm (0.004 in)
Praseodymium

Purity: 99.9% CAS No.: 7440-10-0 Physical Form: ≈0.42g/25x25mm, Packaged in mylar under argon Odour: Odourless UN No.: 3178 MDL No.: MFCD00011174

Formel: Pr
Molecular Weight: 140.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011174
CAS-Nummer: 7440-10-0
EINECS: 231-120-3
UN: 3178
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR