Praseodym 99,1 Metals basis exTa, rods, Ø 6.35 mm (0.25 in)

Lieferant: Thermo Fisher Scientific

40295.DH 40295.DJ 40295.DM 40295.HF
40295.DHEA 240 EUR
40295.DH 40295.DJ 40295.DM 40295.HF
Praseodym 99,1 Metals basis exTa, rods, Ø 6.35 mm (0.25 in)
Praseodymium

Purity: 99.1% CAS No.: 7440-10-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.22g/mm, Cast rod, diameter is approximate Odour: Odourless

Formel: Pr
Molecular Weight: 140.91 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011174
CAS-Nummer: 7440-10-0
EINECS: 231-120-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR