Flußsäure 38,5 - 41,5%
Lieferant: Honeywell Chemicals
|
Gefahr
|
01071.9030
01071.91000
HONE01071.9030EA
0
EUR
HONE01071.9030
HONE01071.91000
Flußsäure 38,5 - 41,5%
Hydrofluoric acid
Formel:
HF Molecular Weight: 20.01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011346 CAS-Nummer: 7664-39-3 EINECS: 231-634-8 UN: 1790 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...