(±)-1,2-Dibromhexafluorpropan
Lieferant: FLUOROCHEM
003478-25G
003478-100G
FLUO003478-25GEA
82.1
EUR
FLUO003478-25G
FLUO003478-100G
(±)-1,2-Dibromhexafluorpropan
(±)-1,2-Dibromhexafluorpropan
Formel:
BrF₂CBrCFCF₃ Molecular Weight: 309,83 g/mol |
MDL:
MFCD00013568 CAS-Nummer: 661-95-0 EINECS: 211-550-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...