(±)-1,1,1-Trichlor-2-propanol

Lieferant: CHEMPUR

RJC03727.250 RJC03727.1
CHMPRJC03727.250EA 0 EUR
CHMPRJC03727.250 MAYBRJC03727.1
(±)-1,1,1-Trichlor-2-propanol
(±)-1,1,1-Trichlor-2-propanol
Formel: Cl₃CCHOHCH₃
Molecular Weight: 163,43 g/mol
CAS-Nummer: 76-00-6
EINECS: 200-924-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR