(±)-2-Hexanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-Butyl-methylcarbinol, (±)-Methyl-butylcarbinol, (±)-2-Hydroxyhexan

128570-100G 128570-25G
SIAL128570-100GEA 296 EUR
SIAL128570-100G SIAL128570-25G
(±)-2-Hexanol, Sigma-Aldrich®
(±)-2-Hexanol
Formel: C₆H₁₄O
Molecular Weight: 102,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004585
CAS-Nummer: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db7ec887376d519b077785cdead15c1aed20d58&vid=5c0c76fb523ec80ddb5134157d5d19ce7a63dcc3&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR