(±)-2-Pentanol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(±)-Methyl-propylcarbinol, (±)-2-Hydroxypentan, (±)-Propyl-methylcarbinol, DL-2-Pentanol, Methylpropylcarbinol
129980010
129985000
129980025
ACRO129980010EA
105
EUR
ACRO129980010
ACRO129985000
ACRO129980025
(±)-2-Pentanol 98%
(±)-2-Pentanol
Formel:
C₅H₁₂O Molecular Weight: 88,15 g/mol Siedepunkt: 118…119 °C (760 mmHg) Schmelzpunkt: –50 °C Dichte: 0,812 g/cm³ Flammpunkt: 34 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004579 CAS-Nummer: 6032-29-7 EINECS: 227-907-6 UN: 1105 ADR: 3,III Merck Index: 12,07258 |
Spezifikation Testergebnisse
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4050 to 1.4070 (20°C, 589 nm) |
Water | ≤0.3 % |
GC | ≥97.5 % |
Residue after evaporation | ≤0.003 % |
Acidity (CH3COOH) | ≤0.005 % |
Ketone | ≤1 % (as methyl propyl ketone) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...