(±)-2-Pentanol ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (±)-Methyl-propylcarbinol, (±)-2-Hydroxypentan, (±)-Propyl-methylcarbinol, DL-2-Pentanol, Methylpropylcarbinol

B21217.0F B21217.AP
B21217.0FEA 235 EUR
B21217.0F B21217.AP
(±)-2-Pentanol ≥99%
(±)-2-Pentanol

Purity: 99% CAS No.: 6032-29-7 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 33°(91°F) Physical Form: Liquid Density: 0.811 Refractive Index: 1.4060 Odour: Characteristic Melting Point: -50° Boiling Point: 114-115° Beilstein Reference: 1718819 Merck Reference: 14,7119 UN No.: 1105 MDL No.: MFCD00004579

Formel: C₅H₁₂O
Molecular Weight: 88,15 g/mol
Siedepunkt: 114…115 °C
Schmelzpunkt: –50 °C
Dichte: 0,811
Flammpunkt: 33 °C (91 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004579
CAS-Nummer: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3,III
Merck Index: 14,7119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc37771666c375a7768be6819fbc64e80617a202&vid=cc73accd09f3b4e43eedad1fe603cf52c119ff25&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR