(±)-3-Octanol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DL-3-Octanol, (±)-Pentyl-ethylcarbinol, Ethylpentylcarbinol, (±)-3-Hydroxyoctan, (±)-Ethyl-pentylcarbinol
A14488.18
A14488.30
A14488.18EA
64.5
EUR
A14488.18
A14488.30
(±)-3-Octanol ≥97%
(±)-3-Octanol
Purity: 97%
CAS No.: 589-98-0
Molecular Formula: C8H18O
Formula Weight: 130.23
Flash Point: 68°(154°F)
Refractive Index: 1.4270
Odour: Pleasant
Melting Point: -45°
Boiling Point: 175-176°
Beilstein Reference: 1719310
MDL No.: MFCD00004590
Formel:
C₈H₁₈O Siedepunkt: 175…176 °C Schmelzpunkt: –45 °C Dichte: 0,820 Flammpunkt: 68 °C (154 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004590 CAS-Nummer: 589-98-0 EINECS: 209-667-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...