DL-Kampfer 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanon, (±)-1,7,7-Trimethyl-2-norbornanon, (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanon

A10936.0C A10936.0E A10936.22 A10936.36
A10936.0CEA 392 EUR
A10936.0C A10936.0E A10936.22 A10936.36
DL-Kampfer 96%
DL-Kampfer

Purity: 96% CAS No.: 76-22-2 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 64°(147°F) Physical Form: Crystalline powder or lumps Density: 0.992 Appearance: White Odour: Camphor-like Melting Point: 172-176° Boiling Point: 204° Solubility: Soluble in alcohols, ether, chloroform, benzene Beilstein Reference: 1907611 Merck Reference: 14,1732 UN No.: 2717 MDL No.: MFCD00074738

Formel: C₁₀H₁₆O
Molecular Weight: 152.24 g/mol
Siedepunkt: 204 °C
Schmelzpunkt: 172…176 °C
Dichte: 0.992
Flammpunkt: 64 °C (147 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00074738
CAS-Nummer: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR