Natriumthiosulfat concentrate for dilution to 0.1 N aqueous solution, Acculute™ Standard Volumetric Solution
Lieferant: Thermo Fisher Scientific
Synonyme: di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit
88628.LQ 88628.LS
88628.LQEA 73.1 EUR
88628.LQ 88628.LS
Natriumthiosulfat concentrate for dilution to 0.1 N aqueous solution, Acculute™ Standard Volumetric Solution
Natriumthiosulfat
CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid MDL No.: MFCD00003499
Formel: Na₂S₂O₃
Molecular Weight: 158.11 g/mol
Siedepunkt: 100
Schmelzpunkt: 48
Lagertemperatur: Raumtemperatur
MDL: MFCD00003499
CAS-Nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844