(S)-(+)-Epichlorhydrin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (S)-(+)-3-Chlor-1,2-epoxypropan, (S)-(+)-Glycerolepichlorhydrin, (S)-(+)-1-Chlor-2,3-epoxypropan

L14298.03 L14298.06 L14298.14
L14298.03EA 55 EUR
L14298.03 L14298.06 L14298.14
(S)-(+)-Epichlorhydrin ≥98%
(S)-(+)-Epichlorhydrin

Purity: 98+% CAS No.: 67843-74-7 Molecular Formula: C3H5ClO Formula Weight: 92.53 Storage Temperature: 2°C to 8°C Flash Point: 32°(90°F) Refractive Index: 1.4380 Boiling Point: 116-117° Beilstein Reference: 1420784 Optical Rotation: +35° (c=1 in methanol) UN No.: 2023 MDL No.: MFCD00077760

Formel: ClCH₂CH(O)CH₂
Molecular Weight: 92,52 g/mol
Siedepunkt: 116…117 °C
Schmelzpunkt: –48 °C
Dichte: 1,180
Flammpunkt: 32 °C (90 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00077760
CAS-Nummer: 67843-74-7
UN: 2023
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR