L(-)-Fenchon 98+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(1R,4S)-(-)-1,3,3-Trimethylbicyclo[2.2.1]-2-heptanon, (1R)-(-)-1,3,3-Trimethylbicyclo[2.2.1]-2-heptanon
188110500
188112500
ACRO188110500EA
34.9
EUR
ACRO188110500
ACRO188112500
L(-)-Fenchon 98+%
L(-)-Fenchon
Formel:
C₁₀H₁₆O Molecular Weight: 152.24 g/mol Siedepunkt: 192 Schmelzpunkt: 5 °C Dichte: 0.94 g/cm³ (0 °C) Flammpunkt: 52 °C |
MDL:
MFCD00151104 CAS-Nummer: 7787-20-4 EINECS: 232-107-5 UN: 1224 ADR: 3 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4600 to 1.4630 (20°C, 589 nm) |
GC | ≥98.0 % |
Specific optical rotation | -47.5° to -53.5° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...