L(-)-Fenchon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (1R,4S)-(-)-1,3,3-Trimethylbicyclo[2.2.1]-2-heptanon, (1R)-(-)-1,3,3-Trimethylbicyclo[2.2.1]-2-heptanon

L15151.18 L15151.30
L15151.18EA 31.4 EUR
L15151.18 L15151.30
L(-)-Fenchon ≥98%
L(-)-Fenchon

Purity: 98+% CAS No.: 7787-20-4 Molecular Formula: C10H16O Formula Weight: 152.24 Flash Point: 73°(163°F) Refractive Index: 1.4630 Melting Point: 3-5° Boiling Point: 192-194° Beilstein Reference: 2042710 Optical Rotation: -52° (neat) MDL No.: MFCD00151104

Formel: C₁₀H₁₆O
Molecular Weight: 152.24 g/mol
Siedepunkt: 192…194 °C
Schmelzpunkt: 3…5 °C
Dichte: 0.945
Flammpunkt: 73 °C (163 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151104
CAS-Nummer: 7787-20-4
EINECS: 232-107-5
UN: 2717
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR