(S)-(-)-Limonen ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-(-)-p-Mentha-1,8-dien, (S)-(-)-Dipenten, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexen, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexen, L(-)-Limonen
L13244.18
L13244.30
L13244.18EA
27.4
EUR
L13244.18
L13244.30
(S)-(-)-Limonen ≥97%
(S)-(-)-Limonen
Purity: 97%
CAS No.: 5989-54-8
Molecular Formula: C10H16
Formula Weight: 136.23
Flash Point: 48°(118°F)
Refractive Index: 1.4720
Melting Point: -74°
Boiling Point: 175-177°
Beilstein Reference: 2323991
Merck Reference: 14,5493
Optical Rotation: -90° (c=10 in ethanol)
UN No.: 2052
MDL No.: MFCD00001558
Formel:
C₁₀H₁₆ Siedepunkt: 175…177 °C Schmelzpunkt: –74 °C Dichte: 0,844 Flammpunkt: 48 °C (118 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001558 CAS-Nummer: 5989-54-8 EINECS: 227-815-6 UN: 2052 ADR: 3,III Merck Index: 14,5493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...