Natriumdichromat Dihydrat 99,5-100,5% ACS

Lieferant: Thermo Scientific
Gefahr

36692.22 36692.36 36692.A3 36692.PG
36692.22EA 40.7 EUR
36692.22 36692.36 36692.A3 36692.PG
Natriumdichromat Dihydrat 99,5-100,5% ACS
Natriumdichromat Dihydrat

Purity: 99.5-100.5% CAS No.: 7789-12-0 Molecular Formula: Cr2Na2O7.2H2O Formula Weight: 298.00 (261.97anhy) Physical Form: Crystalline Density: 2.348 Appearance: Red Odour: Odourless Melting Point: 100° -2H2O Merck Reference: 14,8609 UN No.: 3087 MDL No.: MFCD00149166

Formel: Na₂Cr₂O₇·2H₂O
Molecular Weight: 298,00 g/mol (261,97 anhy)
Schmelzpunkt: 100 °C (-2H₂O)
Dichte: 2,348
Lagertemperatur: Raumtemperatur
MDL: MFCD00149166
CAS-Nummer: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II
Merck Index: 14,8609

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR