Natriumdihydrogenphosphat Monohydrat 90,0-102,0 Alkalimetric, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

Lieferant: Merck

Synonyme: Natriumphosphat monobasisch Monohydrat

MQ300

1.06346.0500 1.06346.1000 1.06346.9025 1.06346.9050 1.06346.9012
1.06346.0500EA 36.17 EUR
1.06346.0500 1.06346.1000 1.06346.9025 1.06346.9050 1.06346.9012
Natriumdihydrogenphosphat Monohydrat 90,0-102,0 Alkalimetric, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Molecular Weight: 137.99 g/mol
Schmelzpunkt: 100
Dichte: 2.04 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (Alkalimetric) 99.0 - 102.0%
Insoluble Matter ≤0.01%
pH-Value (5%; Water, 25 °C) 4.1 - 4.5
UV Absorption (280 nm; 1 mol/L; 1 cm; Water) ≤0.05
Chloride (Cl) ≤0.0005%
Sulfate (SO₄) ≤0.003%
Total Nitrogen (N) ≤0.001%
Heavy Metals (ACS) ≤0.0005%
Ca (Calcium) ≤0.005%
Fe (Iron) ≤0.0005%
K (Potassium) ≤0.005%
Corresponds to ACS, Reag. Ph. Eur.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR