Natriumthiosulfat Pentahydrat 99,5-101,0% ACS
Lieferant: Thermo Fisher Scientific
Synonyme: Fixiersalz
14518.36 14518.A3
14518.36EA 36 EUR
14518.36 14518.A3
Natriumthiosulfat Pentahydrat 99,5-101,0% ACS
Natriumthiosulfat Pentahydrat
Purity: 99.5-101.0% CAS No.: 10102-17-7 Molecular Formula: Na2O3S2.5H2O Formula Weight: 248.17 (158.10anhy) Physical Form: Crystalline Density: 1.69 Appearance: Colourless Odour: Odourless pH: 6.5-8.0 Melting Point: 48° Boiling Point: 100° -5H2O Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,8694 MDL No.: MFCD00149186
Formel: Na₂S₂O₃.5H₂O
Molecular Weight: 248.19 g/mol
Siedepunkt: 100 °C (–5H₂O)
Schmelzpunkt: 48
Dichte: 1.69
Lagertemperatur: Raumtemperatur
MDL: MFCD00149186
CAS-Nummer: 10102-17-7
EINECS: 231-867-5
Merck Index: 14,8694