2-Iodpropan ≥98% stabilisiert
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Isopropyliodid
A15066.36
A15066.14
A15066.22
A15066.36EA
148
EUR
A15066.36
A15066.14
A15066.22
2-Iodpropan ≥98% stabilisiert
2-Iodpropan
Stabilized with copper
Purity: 98+%
CAS No.: 75-30-9
Molecular Formula: (CH3)2CHI
Formula Weight: 169.99
Flash Point: 42°(107°F)
Physical Form: Liquid
Density: 1.701
Refractive Index: 1.4982
Sensitivity: Light Sensitive
Odour: Recognizable
Melting Point: -90°
Boiling Point: 88-90°
Beilstein Reference: 1098244
Merck Reference: 14,5214
UN No.: 2392
MDL No.: MFCD00001071
Formel:
C₃H₇I Molecular Weight: 169.99 g/mol Siedepunkt: 88…90 °C Schmelzpunkt: –90 °C Dichte: 1.701 Flammpunkt: 42 °C (107 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001071 CAS-Nummer: 75-30-9 EINECS: 200-859-3 UN: 2392 ADR: 3 III Merck Index: 14,5214 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...