Zinn(IV)chlorid Pentahydrat 98%, lumps

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Zinntetrachlorid Pentahydrat

11570.22 11570.36
11570.22EA 38.1 EUR
11570.22 11570.36
Zinn(IV)chlorid Pentahydrat 98%, lumps
Zinn(IV)chlorid Pentahydrat

Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapour deposition method.


  • Soluble in water and alcohol


Purity: 98% CAS No.: 7646-78-8 Molecular Formula: Cl4Sn.xH2O Formula Weight: 260.50(anhy) Physical Form: Lump Appearance: Off-white Odour: Pungent Melting Point: 56° Solubility: Soluble in water, alcohol Merck Reference: 14,8774 UN No.: 2440 MDL No.: MFCD00149864

Formel: Cl₄Sn·5H₂O
Molecular Weight: 350.6 g/mol
Schmelzpunkt: 56 °C
Dichte: 2.2 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149864
CAS-Nummer: 10026-06-9
EINECS: 231-588-9
UN: 2440
ADR: 8 III
Merck Index: 14,8774

Bestellen

Spezifikation Testergebnisse

Assay (unspecified) 97.6% min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR