Zink(II)nitrat Hexahydrat ≥99% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Zinknitrat Hexahydrat
12313.A3 12313.36 12313.A9
12313.A3EA 157 EUR
12313.A3 12313.36 12313.A9
Zink(II)nitrat Hexahydrat ≥99% (Metall-Basis)
Zink(II)nitrat Hexahydrat
Purity: 99% CAS No.: 10196-18-6 Molecular Formula: N2O6Zn.6H2O Formula Weight: 297.47 (189.38anhy) Physical Form: Crystalline Density: 2.07 Appearance: White Odour: Odourless Melting Point: ca 36° Solubility: Soluble in water, alcohol Merck Reference: 14,10144 UN No.: 1514 MDL No.: MFCD00149889
Formel: ZnN₂O₆·6H₂O
Molecular Weight: 297,49 g/mol
Schmelzpunkt: ∼36 °C
Dichte: 2,07
Lagertemperatur: Raumtemperatur
MDL: MFCD00149889
CAS-Nummer: 10196-18-6
EINECS: 231-943-8
UN: 1514
ADR: 5.1,II
Merck Index: 14,10144