1,10-Dibromdecan ≥97%

Lieferant: Thermo Scientific

Synonyme: 1,10-Dibromodecan, Decane, 1,10-dibromo-

A12357.22 A12357.36
A12357.22EA 83.5 EUR
A12357.22 A12357.36
1,10-Dibromdecan ≥97%
1,10-Dibromdecan

Purity: 97% CAS No.: 4101-68-2 Molecular Formula: Br(CH2)10Br Formula Weight: 300.08 Flash Point: 110°(230°F) Refractive Index: 1.4930 Melting Point: 26-30° Boiling Point: 157-159°/14mm Beilstein Reference: 506156 MDL No.: MFCD00000222

Formel: BrH₂C(CH₂)₈CH₂Br
Molecular Weight: 300,08 g/mol
Siedepunkt: 157…159 °C (14 mmHg)
Schmelzpunkt: 26…30 °C
Dichte: 1,340
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000222
CAS-Nummer: 4101-68-2
EINECS: 223-871-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR