1-Bromnonan ≥99%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Nonylbromid, 1-Nonylbromid
A10577.14
A10577.22
A10577.36
A10577.14EA
34
EUR
A10577.14
A10577.22
A10577.36
1-Bromnonan ≥99%
1-Bromnonan
Purity: 99%
CAS No.: 693-58-3
Molecular Formula: CH3(CH2)8Br
Formula Weight: 207.16
Flash Point: 90°(194°F)
Refractive Index: 1.4550
Boiling Point: 200-201°
Beilstein Reference: 1733632
MDL No.: MFCD00000278
Formel:
BrH₂C(CH₂)₇CH₃ Molecular Weight: 207,15 g/mol Siedepunkt: 200…201 °C Schmelzpunkt: –29 °C Dichte: 1,086 Flammpunkt: 90 °C (194 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000278 CAS-Nummer: 693-58-3 EINECS: 211-755-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...