1-Bromtetradecan ≥98%

Lieferant: Thermo Scientific

Synonyme: Myristylbromid, Tetradecylbromid

A16082.0E A16082.22 A16082.36
A16082.0EEA 344 EUR
A16082.0E A16082.22 A16082.36
1-Bromtetradecan ≥98%
1-Bromtetradecan

Purity: 98% CAS No.: 112-71-0 Molecular Formula: CH3(CH2)13Br Formula Weight: 277.30 Flash Point: 158°(316°F) Refractive Index: 1.4600 Melting Point: 4-6° Boiling Point: 175-178°/20mm Beilstein Reference: 1742640 MDL No.: MFCD00000228

Formel: BrH₂C(CH₂)₁₂CH₃
Molecular Weight: 277,29 g/mol
Siedepunkt: 175…178 °C (20 mmHg)
Schmelzpunkt: 4…6 °C
Dichte: 1,018
Flammpunkt: 158 °C (316 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000228
CAS-Nummer: 112-71-0
EINECS: 203-999-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR