2-Brombutan ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
sek-Butylbromid
A15069.0B
A15069.30
A15069.0BEA
114
EUR
A15069.0B
A15069.30
2-Brombutan ≥98%
2-Brombutan
Purity: 98%
CAS No.: 78-76-2
Molecular Formula: C4H9Br
Formula Weight: 137.03
Flash Point: 13°(55°F)
Physical Form: Liquid
Density: 1.260
Refractive Index: 1.4369
Appearance: Colourless
Odour: Pleasant
Melting Point: -112°
Boiling Point: 91-92°
Beilstein Reference: 505949
Merck Reference: 14,1554
UN No.: 2339
MDL No.: MFCD00000156
Formel:
H₃CBrCHCH₂CH₃ Molecular Weight: 137,02 g/mol Siedepunkt: 91…92 °C Schmelzpunkt: –112 °C Dichte: 1,260 Flammpunkt: 13 °C (55 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000156 CAS-Nummer: 78-76-2 EINECS: 201-140-7 UN: 2339 ADR: 3,II Merck Index: 14,1554 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...