2-Brompropan ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
iso-Propylbromid
A12944.AK
A12944.AU
A12944.AKEA
46.5
EUR
A12944.AK
A12944.AU
2-Brompropan ≥99%
2-Brompropan
Purity: 99%
CAS No.: 75-26-3
Molecular Formula: C3H7Br
Formula Weight: 123.00
Flash Point: 19°(66°F)
Physical Form: Liquid
Density: 1.310
Refractive Index: 1.4255
Appearance: Colourless to pale yellow
Melting Point: -89°
Boiling Point: 59-60°
Beilstein Reference: 741852
Merck Reference: 14,5210
UN No.: 2344
MDL No.: MFCD00000147
Formel:
H₃CBrCHCH₃ Molecular Weight: 122,99 g/mol Siedepunkt: 59…60 °C Schmelzpunkt: –89 °C Dichte: 1,310 Flammpunkt: 19 °C (66 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000147 CAS-Nummer: 75-26-3 EINECS: 200-855-1 UN: 2344 ADR: 3,II Merck Index: 14,5210 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...