4-Bromheptan 97%

Lieferant: Thermo Fisher Scientific
Achtung

L04807.06 L04807.14
L04807.06EA 51.8 EUR
L04807.06 L04807.14
4-Bromheptan 97%
4-Bromheptan

Purity: 97% CAS No.: 998-93-6 Molecular Formula: C7H15Br Formula Weight: 179.11 Flash Point: 40°(104°F) Refractive Index: 1.4500 Boiling Point: 60-62°/18mm Beilstein Reference: 1697144 UN No.: 1993 MDL No.: MFCD00039182

Formel: H₃C(CH₂)₂BrCH(CH₂)₂CH₃
Molecular Weight: 179.1 g/mol
Siedepunkt: 60…62 °C (18 mmHg)
Dichte: 1.140
Flammpunkt: 40 °C (104 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039182
CAS-Nummer: 998-93-6
EINECS: 213-653-3
UN: 1993
ADR: 3 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR