1,2-Dibrom-1-chlor-1,2,2-trifluorethan

Lieferant: CHEMPUR

Synonyme: 1-Chlor-1,2-dibrom-1,2,2-trifluorethan

BTB11038.250 BTB11038.1
CHMPBTB11038.250EA 0 EUR
CHMPBTB11038.250 MAYBBTB11038.1
1,2-Dibrom-1-chlor-1,2,2-trifluorethan
1,2-Dibrom-1-chlor-1,2,2-trifluorethan
Formel: BrClFCCBrF₂
Molecular Weight: 276,28 g/mol
MDL: MFCD00039316
CAS-Nummer: 354-51-8
EINECS: 206-562-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR