1,4-Dibrom-1,1,2,2-tetrafluorbutan

Lieferant: FLUOROCHEM

010444-25G 010444-5G
FLUO010444-25GEA 0 EUR
FLUO010444-25G FLUO010444-5G
1,4-Dibrom-1,1,2,2-tetrafluorbutan
1,4-Dibromo-1,1,2,2-tetrafluorobutane
Formel: BrF₂CCF₂CH₂CH₂Br
MDL: MFCD00039224
CAS-Nummer: 18599-20-7
EINECS: 242-438-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR