Propionaldehyd ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Propanal

A16146.0J A16146.AP A16146.AU
A16146.0JEA 162 EUR
A16146.0J A16146.AP A16146.AU
Propionaldehyd ≥97%
Propionaldehyd

Purity: 97% CAS No.: 123-38-6 Molecular Formula: CH3CH2CHO Formula Weight: 58.08 Storage Temperature: 2°C to 8°C Flash Point: -40°(-40°F) Refractive Index: 1.3620 Odour: Pungent Melting Point: -81° Boiling Point: 47-49° Beilstein Reference: 506010 Merck Reference: 14,7823 UN No.: 1275 MDL No.: MFCD00007020

Formel: C₃H₆O
Molecular Weight: 58,08 g/mol
Siedepunkt: 47…49 °C
Schmelzpunkt: –81 °C
Dichte: 0,81
Flammpunkt: –40 °C (–40 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00007020
CAS-Nummer: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 14,7823

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR