(1-Dodecyl)triphenylphosphoniumbromid

Lieferant: CHEMPUR
Achtung

Synonyme: Dodecyltriphenylphosphonium bromide

BTB13910.250 BTB13910.1
CHMPBTB13910.250EA 0 EUR
CHMPBTB13910.250 MAYBBTB13910.1
(1-Dodecyl)triphenylphosphoniumbromid
(1-Dodecyl)triphenylphosphoniumbromid
Formel: H₃C(CH₂)₁₁P(C₆H₅)₃Br
Molecular Weight: 511.53 g/mol
MDL: MFCD00031546
CAS-Nummer: 15510-55-1
EINECS: 239-538-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0fe7db87e7b9e3a4f94baa8840868d0edbeaaa6&vid=e0fe7db87e7b9e3a4f94baa8840868d0edbeaaa6&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR