L(+)-Isoleucin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
(2S,3S)-(+)-2-Amino-3-methylpentansäure, (2S,3S)-(+)-α-Amino-β-methylvaleriansäure, (2S,3S)-(+)-Isoleucin
4030756.0100
4030756.0500
BACE4030756.0100EA
0
EUR
BACE4030756.0100
BACE4030756.0500
L(+)-Isoleucin
L(+)-Isoleucin
Sequence: H-Ile-OH
Formel:
H₃CCH₂CH(CH₃)CH(NH₂)COOH Molecular Weight: 131.17 g/mol Schmelzpunkt: 279…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064222 CAS-Nummer: 73-32-5 EINECS: 200-798-2 Merck Index: 14,05179 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...