1,1,1-Trifluor-3-phenylaceton
Lieferant: FLUOROCHEM
Synonyme:
3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one
CFF436.5
CFF436.10
CFF436.25
017451-5G
017451-10G
017451-25G
CHMPCFF436.5EA
348
EUR
CHMPCFF436.5
CHMPCFF436.10
CHMPCFF436.25
FLUO017451-5G
FLUO017451-10G
FLUO017451-25G
1,1,1-Trifluor-3-phenylaceton
1,1,1-Trifluor-3-phenylaceton
Formel:
C₉H₇F₃O Molecular Weight: 188.15 g/mol |
MDL:
MFCD00068171 CAS-Nummer: 350-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...