(3,3,3-Trifluorpropyl)trimethoxysilan

Lieferant: CHEMPUR SILANE

Synonyme: Trimethoxy(3,3,3-trifluorpropyl)silan

SILS17850.5 SILS17850.25
SILNSILS17850.5EA 0 EUR
SILNSILS17850.5 SILNSILS17850.25
(3,3,3-Trifluorpropyl)trimethoxysilan
(3,3,3-Trifluoropropyl)trimethoxysilane
Formel: F₃CCH₂CH₂Si(OCH₃)₃
Molecular Weight: 218.25 g/mol
Siedepunkt: 144
Dichte: 1.137 g/cm³ (20 °C)
MDL: MFCD00039266
CAS-Nummer: 429-60-7
EINECS: 207-059-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR