3-Phenylpropylamin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Amino-3-phenylpropane, 1-Amino-3-phenylpropan

L03023.09 L03023.18
L03023.09EA 30.2 EUR
L03023.09 L03023.18
3-Phenylpropylamin ≥98%
3-Phenylpropylamin

Purity: 98+% CAS No.: 2038-57-5 Molecular Formula: C6H5CH2CH2CH2NH2 Formula Weight: 135.21 Flash Point: 90°(194°F) Refractive Index: 1.5250 Odour: Amine-like Boiling Point: 220-221° Beilstein Reference: 2205526 UN No.: 2735 MDL No.: MFCD00008224

Formel: C₉H₁₃N
Molecular Weight: 135,21 g/mol
Siedepunkt: 220…221 °C
Dichte: 0,949
Flammpunkt: 90 °C (194 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008224
CAS-Nummer: 2038-57-5
EINECS: 218-012-1
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR