3-Phenylpropylamin

Lieferant: CHEMPUR

Synonyme: 1-Amino-3-phenylpropane, 1-Amino-3-phenylpropan

JFD03595.250 JFD03595.1
CHMPJFD03595.250EA 0 EUR
CHMPJFD03595.250 MAYBJFD03595.1
3-Phenylpropylamin
3-Phenylpropylamin
Formel: C₉H₁₃N
Molecular Weight: 135.21 g/mol
Siedepunkt: 221
Dichte: 0.951
MDL: MFCD00008224
CAS-Nummer: 2038-57-5
EINECS: 218-012-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR