(3-Bromphenyl)essigsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-(3-Bromophenyl)acetic acid
CA-4261-25G
CA-4261-100G
COBBCA-4261-25GEA
61.3
EUR
COBBCA-4261-25G
COBBCA-4261-100G
(3-Bromphenyl)essigsäure
(3-Bromphenyl)essigsäure
Formel:
(C₆H₄Br)CH₂COOH Molecular Weight: 215,05 g/mol Siedepunkt: 308 °C (1013 hPa) Schmelzpunkt: 102…103 °C |
MDL:
MFCD00004330 CAS-Nummer: 1878-67-7 EINECS: 217-522-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...