(3-Methoxyphenyl)essigsäureethylester ≥98%

Lieferant: Thermo Scientific

Synonyme: Ethyl-(3-methoxyphenyl)acetat

A15070.06 A15070.14
A15070.06EA 130 EUR
A15070.06 A15070.14
(3-Methoxyphenyl)essigsäureethylester ≥98%
(3-Methoxyphenyl)essigsäureethylester

Purity: 98% CAS No.: 35553-92-5 Molecular Formula: C11H4O3 Formula Weight: 194.23 Boiling Point: 109-110°/1mm MDL No.: MFCD00026897

Formel: C₁₁H₁₄O₃
Molecular Weight: 194,23 g/mol
Siedepunkt: 109…110 °C (1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00026897
CAS-Nummer: 35553-92-5
EINECS: 252-614-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR