(3-Pyridyl)essigsäureethylester ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl-(3-pyridyl)acetat

A14768.06 A14768.14
A14768.06EA 34.5 EUR
A14768.06 A14768.14
(3-Pyridyl)essigsäureethylester ≥99%
(3-Pyridyl)essigsäureethylester

Purity: 99% CAS No.: 39931-77-6 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Refractive Index: 1.500 Boiling Point: 79-80°/0.4mm Beilstein Reference: 123827 MDL No.: MFCD00006411

Formel: C₉H₁₁NO₂
Molecular Weight: 165.19 g/mol
Siedepunkt: 79…80 °C (0.4 mmHg)
Dichte: 1.086
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006411
CAS-Nummer: 39931-77-6
EINECS: 254-707-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR