(4-Aminophenyl)essigsäureethylester 98%

Lieferant: Thermo Scientific

Synonyme: Ethyl-(4-aminophenyl)acetat

A11529.06 A11529.14
A11529.06EA 70.8 EUR
A11529.06 A11529.14
(4-Aminophenyl)essigsäureethylester 98%
(4-Aminophenyl)essigsäureethylester

Purity: 98% CAS No.: 5438-70-0 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 46-48° MDL No.: MFCD00017569

Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
Schmelzpunkt: 46…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017569
CAS-Nummer: 5438-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR